Forceps, Gemologist, Starlight Coated Tip, Excelta Corp®
Supplier: Excelta
JLS-4 JS-1 JS-10 JS-11 JSS-1
75880-468EA 124.99 CAD
75880-468 75880-476 75880-750 75880-752 75880-754
Forceps, Gemologist, Starlight Coated Tip, Excelta Corp®
Forceps
Designed for handling gem stones.
  • Starlight coated tips hold gems more securely
  • Various sizes available
  • Stainless steel corrosion resistant
These gemologist straight medium point tweezers are ideal for handling medium gems. The handles are serrated for better control. They feature 'starlight' coated tips.