Forceps, Gemologist, Longitudinal Groove, Slide Lock, Excelta Corp®
Supplier: Excelta
JLS-5 JLB-2 JLB-4 JLS-2
75880-470EA 128.31 CAD
75880-470 75880-736 75880-740 75880-744
Forceps, Gemologist, Longitudinal Groove, Slide Lock, Excelta Corp®
Forceps
Designed for handling gem stones - Groove and Lock hold gem stones securely.
  • Longitudinal groove hold gem securely
  • Slide lock allows gem to be held indefinitly
  • Various sizes available
  • Stainless steel corrosion resistant
These gemologist straight medium point tweezers are ideal for handling medium gems. The handles are serrated for better control. They feature both a 'slide-lock' and a 'longitudinal groove in the tips' to both seat the gem in a secure fashion and lock the gem in place.