Tweezers, Super Neverust High Precision Tips, Excelta Corp®
Supplier: Excelta
2A-SN 3-SN 3C-SN 5-SN 7-SN
75880-770EA 161.32 CAD
75880-770 75880-774 75880-784 75880-790 75880-798
Tweezers, Super Neverust High Precision Tips, Excelta Corp®
Forceps
For cleanroom applications.
  • Cleanroom safe, (AISI 904l) stainless steel
  • Available in several tip configurations
  • Very precise tips
These four star precision pointed tweezer are made of AISI (904L) Super Neverust®, which has excellent mechanical strength, good surface stability and high resistance to corrosion or oxidation.
Order Now
SPECIFICATIONS
Environmentally Preferable