Tweezers, Diamond Coated Tips, Excelta Corp®
Supplier: Excelta
SS-SA-DC 2A-SA-DC 00-SA-DC 3C-SA-DC
75880-504EA 182.06 CAD
75880-504 75880-522 75880-764 75880-778 75880-788 75880-796
Tweezers, Diamond Coated Tips, Excelta Corp®
Forceps
Designed for handling SMD components - Coated tips add strength and longevity to the tips.
  • Biocompatible
  • Chemically inert to 350 °C
  • Prevents particle shedding
  • Cleanroom safe, stainless steel
  • Protects the tips from wear while providing elasticity
These unique 'Diamond Coated' high precision pointed tweezers are made of anti-magnetic Neverust® stainless steel. The diamond coated tips are biocompatible, chemically inert to 350°C, prevent particle shedding, cleanroom safe, and protect the tips from wear while providing elasticity.