VWR® LTS Compatible Pipette Tips
Supplier: VWR International
76322-158 76322-160 76322-162 76322-164 76322-172 76322-174 76322-176 76322-178 76322-180 76322-530 76322-532 76322-534 76323-932 76323-934 76323-936 76323-938 76323-940 76323-942 76323-944 76323-946 76327-210 76327-212 76327-216 76327-218 76327-220
76322-158CS 931.31 CAD
76322-158 76322-160 76322-162 76322-164 76322-172 76322-174 76322-176 76322-178 76322-180 76322-530 76322-532 76322-534 76323-932 76323-934 76323-936 76323-938 76323-940 76323-942 76323-944 76323-946 76327-210 76327-212 76327-216 76327-218 76327-220
VWR® LTS Compatible Pipette Tips
Pipette Tips
VWR® LTS™ Compatible pipette tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS™ compatible pipettors.
  • Compatible with the LTS™ style pipettors
  • Maximizes sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 15 psi, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
Certifications: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.