Level Sensor Adapters, for Bioreactors, Chemglass
Supplier: Chemglass
CLS-1380-92
76333-454EA 445.09 CAD
76333-454
Level Sensor Adapters, for Bioreactors, Chemglass
Fasteners Screws
Used to install a ¹/₄” OØ (6.35 mm) level sensor into an M10 headplate port.