O-Rings, PERFLUORO Perfluoroelastomer Compound, Staver Hydraulics
Supplier: Chemglass
CG-309-015 CG-309-021 CG-309-113 CG-309-213 CG-309-224
76333-408EA 54.76 CAD
76333-408 76333-410 76333-412 76333-414 76333-416 76333-418 76333-420
O-Rings, PERFLUORO Perfluoroelastomer Compound, Staver Hydraulics
Seals Sealing Rings
One of the most thermally stable elastomeric materials available, Perfluoro is a family of perfluoroelastomer compounds designed to provide superior chemical resistance, and they offer a cost savings over similar o-rings.
Perfluoro o-rings are stocked in WHITE 75 durometer (unless otherwise noted). The effective service temperature is −7 to 230 °C. Recommended for use with acids, inorganic bases, alcohols/ethers, aromatic hydro-carbons, esters, aldehydes/ ketones, aliphatic hydrocarbons, amines and chlorocarbon solvents.
Ordering information: For complete chemical resistance and fluid compatibility, contact your Avantor representative.