Stirrer Shaft Couplings, Universal, 100 L Process Reactors, Split Couplings, Chemglass
Supplier: Chemglass
CG-2046-X-130 CG-2046-X-131 CG-2046-X-132
76333-694EA 930.77 CAD
76333-694 76333-696 76333-698
Stirrer Shaft Couplings, Universal, 100 L Process Reactors, Split Couplings, Chemglass
Stirrers Stirring Tools
Split coupling design allows for easier assembly and disassembly between the motor and 30 mm glass stirrer shaft.
Coupling with universal joint compensates for misalignment of drive motor and glass stir shaft. Nylon bushing compresses around shaft using an allen screw. Stainless steel collar/bushing and upper coupling attach directly to the motor shaft. For use with 100 L process reactor systems. 
Order Now
SPECIFICATIONS
Environmentally Preferable