TRU-STIR Stirrer Shaft Couplings, Compact 3" Long, Chemglass
Supplier: Chemglass
CG-2044-C-11 CG-2044-C-12 CG-2044-C-14 CG-2044-C-16 CG-2044-C-10
76333-398EA 590.12 CAD
76333-398 76333-400 76333-402 76333-404 76333-690
TRU-STIR Stirrer Shaft Couplings, Compact 3" Long, Chemglass
Stirrers Stirring Tools
The unique design of our Tru-Stir™ stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to ¹/₈" of offset misalignment. Coupling is rated for 21 in-lbs of torque without the backlash normally associated with common coil springs.
Similar to CG-2044 but the overall height is 1" shorter. The unique design of our Tru-Stir™ stirrer shaft coupling eliminates the need to use rubber hose or universal adapters to attach your stirrer shafts to overhead stir motors. Coupling incorporates a unique helical S.S. spring permitting up to ¹/₈" of offset misalignment. Coupling is rated for 21 in-lbs of torque without the backlash normally associated with common coil springs. Lower portion of coupling has a nylon adapter with a one piece collar for attachment to 10, 19mm stirrer shafts. Upper adapters permit the coupling to be used with a standard Jacobs style 3-jaw chuck or motors having shaft diameters of ⁵/₁₆", ³/₈", 12 mm, ¹/₂", ⁵/₈" or ³/₄". Adapter is also available for use with CG-2047 flexible shafts having ⁵/₁₆" end fittings. Overall length of the coupling is 3".
Delivery information: Supplied complete with an allen wrench set.