Offset Adapter, for Velp Hotplates, Chemglass
Supplier: Chemglass
CG-1995-V-29
76333-664EA 88.79 CAD
76333-664
Offset Adapter, for Velp Hotplates, Chemglass
Fasteners Screws
Offset Adapter, for Velp Hotplates, Stainless steel
This stainless steel offset adapter allows use of the CG-1995-V-22 vertical support rod with larger diameter accessories on CG-1994-V, CG-1995-V, and CG-1997-V series hotplates. Offset adapter can be used with cooling/heating blocks up approx. 8 to 5/8"/220 mm diameter. Offset created by adapter is 20 mm center to center.
Delivery information: Supplied complete with M8 stainless steel bolt.