Spray Ball, Fluid Driven,³/₈" Compression Fitting, PTFE, Chemglass
Supplier: Chemglass
CG-1989-X-22 CG-1989-X-35 CG-1989-X-75
76333-394EA 585.5 CAD
76333-394 76333-396 76333-652
Spray Ball, Fluid Driven,³/₈" Compression Fitting, PTFE, Chemglass
Spray Guns
This fluid driven spray ball gives nearly 360° spray coverage and is a convenient way to clean Chemglass 10 through 100 L glass reactors without dismantling the entire setup. 
  • Spray ball/head - has larger slots for improved spray pattern and velocity of spray
  • Large bore for more flow
  • ³/₈" compression fitting
  • All PTFE body and spray ball
It can also be used as a wall wetter. Spray ball is constructed of PTFE, the removable PTFE clip allows the rotating spray head to be easily removed for inspection and maintenance. ³/₈" top compressionMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable