Air Operated Lower Drain Valves for 10 L Thru 50 L Reactors, Air Actuated, Chemglass
Supplier: Chemglass
CG-1968-V-01 CG-1968-V-02
76333-494EA 10217.24 CAD
76333-494 76333-496
Air Operated Lower Drain Valves for 10 L Thru 50 L Reactors, Air Actuated, Chemglass
Valves Line Valves
This air operated, flush bottom drain valve fits our reactors from 10 to 50 liters that have a 2" beaded pipe connection.
  • Factory preset regulators
  • Valve is operated by turning the handle to the open and close positions
  • Valkve opens and closes quickly and rasily
  • Panel can be mounted outside of hood
This air operated, flush bottom drain valve fits our reactors from 10 to 50 liters that have a 2" beaded pipe connection. This valve is perfect for applications where solvent extractions or partitioning are done after a reaction. Due to the constant springMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable