Drain Valve, Zero Dead Space GL Heavy Duty Threads, Pilot Plant, 2" Beaded Pipe, Torque Knob, Chemglass
Supplier: Chemglass
CG-1968-GL-241
76333-490EA 3030.52 CAD
76333-490
Drain Valve, Zero Dead Space GL Heavy Duty Threads, Pilot Plant, 2" Beaded Pipe, Torque Knob, Chemglass
Valves Line Valves
The “Zero Dead Space” flush bottom valve is designed to prevent solids from collecting in the bottom of the vessel.
  • Glass body features a heavy duty GL style glass thread
  • Auto-stop when opening the valve
  • Anodized aluminum control knob is laser etched with directional arrows for opening and closing the valve
  • Zero dead space
  • Perfluoro O-rings on tip and plunger
The CG-1968-GL valve is compatible for use on all Chemglass reactors that previously used CG-1968-Q series 2" drain valves. The “Zero Dead Space” flush bottom valve is designed to prevent solids from collecting in the bottom of the vessel. The PTFE drain valve plunger is supplied with a Perfluoro and a TFE encapsulated o-ring for the tip and two Perfluoro o-rings installed on the shaft. The drain valve is connected to the reactor by a unique 2" beaded pipe valve seat that is sealed to the reactor bottom. Our valve permits total drainage of the vessel contents.
The sidearm has a 1" beaded pipe fitting. There are two types of plug assemblies; the first is the plain plug for use with our standard agitators; the second has a stabilizing pin that is used with the 100 L agitators. The pin extends into the lower agitator and virtually eliminates any whipping by stabilizing the stirrer shaft.