Drain Valve, Zero Dead Space, GL Heavy Duty Threads, Pilot Plant, 2" Beaded Pipe, Chemglass
Supplier: Chemglass
CG-1968-GL-040 CG-1968-GL-041
76333-386EA 2522.97 CAD
76333-386 76333-388
Drain Valve, Zero Dead Space, GL Heavy Duty Threads, Pilot Plant, 2" Beaded Pipe, Chemglass
Valves Line Valves
The “Zero Dead Space” flush bottom valve is designed to prevent solids from collecting in the bottom of the vessel.
  • Glass body features a heavy duty GL style glass thread
  • Auto-stop when opening the valve
  • Anodized aluminum control knob is laser etched with directional arrows for opening and closing the valve
  • Zero dead space
  • Perfluoro O-rings on tip and plunger
The PTFE drain valve plunger is supplied with a Perfluoro and a TFE encapsulated o-ring for the tip and two Perfluoro o-rings installed on the shaft. The drain valve is connected to the reactorMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable