Mobile Phase HPLC Cap Systems, Chemglass
Supplier: Chemglass
CG-1167-11 CG-1167-12 CG-1167-13 CG-1167-14
76333-330EA 811.4 CAD
76333-330 76333-332 76333-334 76333-336
Mobile Phase HPLC Cap Systems, Chemglass
Closures Safety Caps and Safety Waste Caps
The new and improved CG-1167 HPLC Cap system provides the safest and most economical way of storing high performance, low pressure liquid chromatography mobile phase, sparging preventing bubbles from entering the system as well as collecting sample waste.
The bottle is blue tinted plastic coated flat bottom, which provides additional safety by containing the glass fragments from accidents and allowing enough time to transfer samples or discarding safely.
Delivery information: Plastic Coated Glass Reservoir (1), 3-Hole Cap Assembly (1),
Combination Inlet/Sparger Filter 2 um (1), CG-1164-N-02 Flangless Nuts for 1.8" Tubing (2), CG-1164-F-02 Flangeless Ferrule for ¹/₈" Tubing (2), CG-1166-10 Tefzel Plug (1), CG-1164-02 ¹/₈" PTFE Tubing n 10' long.