Electronic Cell Stirrer, FireflySci
Supplier: FIREFLYSCI, INC.
P82-110VAC P82-240VAC
76016-694EA 4455.25 CAD
76016-694 76016-696
Electronic Cell Stirrer, FireflySci
Stirrers Stirring Tools
Stirs cell contents while measuring absorbance or fluorescence. Fits all standard spectrophotometers and fluorometers with a 10 mm cuvette holder.
  • Controller and electronic magnetic stirring assembly included
  • Controller is 4.0×2.5×3.0" (including speed controller knob)
  • Available for use with 100/120 or 220/240 VAC wall currents
The sample is mixed by a tiny magnetic stir bar which is placed at the bottom of the cell. A small electronic rotation magnet assembly is placed under the cell in the cell compartment. This raises the cell up 5mm, which allowsMore Product Information