Cuvette Stirring Rods, FireflySci
Supplier: FIREFLYSCI, INC.
P67 P68 P69
76016-684EA 87.99 CAD
76016-684 76016-686 76016-688
Cuvette Stirring Rods, FireflySci
Stirrers Stirring Rods
Stirring rods for the manual stirring of samples in a 10 mm cuvette.
  • Steam autoclavable at 121 °C (250 °F)
Stirring rods allow samples to be continuously spun at the bottom of a cuvette to prevent sedimentation and coagulation.