Avantor® ACE® HILIC-A, Analytical, HPLC/UHPLC Columns
Supplier: VWR
Avantor® ACE®
HILA-3-1246U HILA-5-2502U HILA-3-0103GD HILA-5-0520 HILA-3-3502U HILA-5-2520 HILA-3-0502U HILA-3-1546U HILA-17-1002U HILA-5-0502U HILA-5-0303U HILA-5-1520 HILA-17-0503U HILA-3-1046U HILA-5-3503U HILA-5-0302U HILA-5-3546U HILA-5-0103GD HILA-5-1202U HILA-3-0503U HILA-5-2510 HILA-3-3546U HILA-3-0102GD HILA-17-0202U HILA-5-1503U HILA-3-1002U HILA-17-0502U HILA-5-1002U HILA-5-3502U HILA-3-1202U HILA-3-7546U HILA-3-0303U HILA-3-1502U HILA-5-7546U HILA-5-0102GD HILA-5-2546U HILA-5-2530 HILA-5-1220 HILA-5-1030 HILA-5-1008 HILA-5-3030 HILA-5-7503U HILA-5-1203U HILA-3-2502U HILA-17-0203U HILA-5-1010 HILA-5-1530 HILA-5-7530 HILA-17-0303U HILA-3-7502U HILA-3-1503U HILA-3-0203U HILA-5-1246U HILA-17-0302U HILA-5-3010 HILA-5-0530 HILA-3-7503U HILA-5-0203U HILA-17-7503U HILA-3-0302U HILA-3-0346U HILA-5-1020 HILA-3-0202U HILA-5-0546U HILA-17-1003U HILA-5-7502U HILA-5-1508 HILA-3-1203U HILA-5-7520 HILA-5-1510 HILA-17-7502U HILA-5-2503U HILA-5-0120GD HILA-3-0246U HILA-5-0503U HILA-5-1003U HILA-17-3502U HILA-17-3503U HILA-5-0346U HILA-5-0246U HILA-5-1046U HILA-3-0546U HILA-3-2503U HILA-5-0508 HILA-5-2508 HILA-5-0110GD HILA-5-0510 HILA-3-2546U HILA-5-1546U HILA-5-3020 HILA-3-1003U HILA-5-0202U HILA-3-3503U HILA-5-1502U
76395-376EA 1208.37 CAD
76395-376 76395-496 76395-254 76395-298 76395-378 76395-498 76395-256 76395-410 76395-372 76395-294 76395-250 76395-492 76395-374 76395-296 76395-450 76395-252 76395-494 76395-538 76395-416 76395-336 76395-534 76395-412 76395-456 76395-258 76395-536 76395-338 76395-458 76395-414 76395-370 76395-490 76395-266 76395-342 76395-268 76395-344 76395-262 76395-264 76395-340 76395-428 76395-308 76395-506 76395-468 76395-500 76395-346 76395-426 76395-348 76395-502 76395-380 76395-310 76395-430 76395-474 76395-312 76395-510 76395-476 76395-432 76395-394 76395-350 76395-470 76395-352 76395-472 76395-516 76395-438 76395-512 76395-236 76395-478 76395-434 76395-514 76395-436 76395-238 76395-508 76395-390 76395-392 76395-270 76395-244 76395-364 76395-246 76395-322 76395-366 76395-240 76395-284 76395-242 76395-362 76395-406 76395-526 76395-408 76395-528 76395-248 76395-368 76395-324 76395-404 76395-448 76395-326 76395-280 76395-282 76395-480
Avantor® ACE® HILIC-A, Analytical, HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® HILIC-A columns have been designed specifically for HILIC applications and are especially useful for the separation of polar analytes. These columns are part of a suit of 3 proprietary HILIC columns (HILIC-A, HILIC-B and HILIC-N) developed to enable chromatographers to achieve reproducible and successful HILIC separations using a systematic approach to method development.
  • Proprietary SIL (silica) acidic character HILIC phase
  • Compatible with all manufacturers HPLC/UHPLC systems
  • Excellent column performance, inertness and robustness
  • Simplified and structured approach to successful HILIC separations
  • Available in a range of particle sizes and column dimensions including UHPLC/HPLC rated hardware formats
Ordering information: HILIC method development kits, HILIC method development guide and flowchart are available on request.