Avantor® ACE® C18, HPLC Columns, 10 µm
Supplier: HICHROM LIMITED
ACE-131-0120GD ACE-131-1510 ACE-131-1539 ACE-131-1546 ACE-131-2046 ACE-131-1004 ACE-131-1020 ACE-131-1046 ACE-131-1204 ACE-131-1246 ACE-131-1503 ACE-131-1504 ACE-131-1508 ACE-131-2503 ACE-131-2504 ACE-131-2510 ACE-131-2520 ACE-131-2546 ACE-131-3004 ACE-131-3008 ACE-131-3039 ACE-131-3046 ACE-131-0103GD ACE-131-0110GD ACE-131-0346
76388-354PK 6931.07 CAD
76388-354 76388-362 76388-376 76388-378 76388-528 76388-530 76388-532 76388-534 76388-536 76388-548 76388-552 76388-726 76388-728 76388-730 76388-442 76388-444 76388-446 76388-452 76388-454 76388-456 76388-496 76388-498 76388-504 76388-506 76388-520 76388-524 76388-526 76388-606 76388-610 76388-612 76388-614 76388-616 76388-618 76388-620 76388-632 76388-634 76388-636 76388-640 76388-642 76388-644 76388-646 76388-700 76388-346 76388-350 76388-396 76388-404 76388-408 76388-414 76388-422 76388-424 76388-426 76388-428 76388-430
Avantor® ACE® C18, HPLC Columns, 10 µm
Chromatography Columns
Avantor® ACE® C18 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.