Avantor® ACE® C4, HPLC Columns, 3 µm
Supplier: Avantor
ACE-113-15003 ACE-113-1502 ACE-113-1503 ACE-113-1546 ACE-113-0502 ACE-113-0503 ACE-113-0546 ACE-113-10003 ACE-113-1002 ACE-113-1003 ACE-113-1046 ACE113005003GD ACE-113-0101GD ACE-113-0102GD ACE-113-0103GD ACE-113-0302
76384-800EA 1670.72 CAD
76384-800 76384-802 76384-804 76384-806 76384-808 76384-810 76384-812 76384-814 76384-816 76384-818 76384-820 76384-822 76384-824 76384-826 76384-828 76384-830 76384-832 76384-834 76384-836 76384-838 76384-840 76384-842 76384-844 76384-850 76384-854 76384-856 76384-858 76384-860 76384-862 76384-864 76384-866 76384-868 76384-870 76384-872 76384-874 76384-876 76384-878 76384-880 76384-882 76384-756 76384-758 76384-760 76384-762 76384-764 76384-766 76384-768 76384-770 76384-772 76384-774 76384-776 76384-778 76384-780 76384-782 76384-784 76384-786 76384-788 76384-790 76384-792 76384-794 76384-796 76384-798 76384-726 76384-728 76384-730 76384-732 76384-734 76384-736 76384-738 76384-740 76384-742 76384-744 76384-746 76384-748 76384-750 76384-752 76384-754
Avantor® ACE® C4, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® C4 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.