Rebel Hybrid Microscope, Upright and Inverted, Discover Echo
Supplier: DISCOVER ECHO INC.
RSTG1001 RHDR0002 RHDR0006 RBLT3000 RSTG3001 RCND3001 RCND3013 RMAG0113
76307-592EA 1488.63 CAD
76307-592 76307-594 76307-596 76376-746 76376-750 76376-752 76376-754 76376-756 76376-762
Rebel Hybrid Microscope, Upright and Inverted, Discover Echo
Microscopes
Rebel is designed around the user to challenge the status quo. Designed from the ground up to improve microscopy.
  • ECHO Pro: powerful yet simple software reimagined to collaboratively view, capture, and share images
  • Ultra high resolution 13" touch display
  • CMOS color camera (8 MP)
  • Plan Achromat and Apochromat options available
  • XY mechanical stage option available
  • Ergonomic biosafety cabinet and adjustable height table options available
Versatility: Hybrid microscope easily converts between upright and inverted configurations. Brightfield and phase contrast modes available. Eyepiece option available as well.
Productivity: Our award-winning touch interface is incredibly easy to use. Eyepieces have been replaced with a 13" iPad with Retina Display to view and capture stunning crystal-clear images.
Rebel True Ergo Table and Rebel Logic + Biosafety cabinet by Labconco bring the microscope stage down flush to the tabletop, setting a new precendent in microscope ergonomics.
Ordering information:
Microscope includes a one-year warranty. Contact your Avantor representative to demo the the newest innovations in microscopy. A Rebel Hybrid Microscope Bundle option is also available (see 76359-898).
Delivery information: Includes microscope body, iPad with locking stand, 8mp CMOS color camera, power cord, microscope cover, and lens cloth.