Avantor® ACE® C18-300, HPLC Columns, 5 µm
Supplier: Avantor
ACE-221-2546 ACE-221-1510 ACE-221-1546 ACE-221-2502 ACE-221-2503 ACE-221-2504 ACE-221-2510 ACE-221-2520 ACE2212546MVK ACE-221-0102GD ACE-221-0103GD ACE-221-0110GD ACE-221-0502 ACE-221-0546 ACE-221-1002 ACE-221-1008 ACE-221-1046 ACE-221-1202 ACE-221-1502 ACE-221-1503 ACE-221-1504 ACE-221-7502 ACE-221-7546
76382-596EA 2382.06 CAD
76382-596 76390-148 76390-280 76390-282 76390-284 76390-286 76390-288 76390-290 76390-292 76390-294 76390-304 76390-306 76390-308 76390-310 76390-312 76390-314 76390-316 76390-318 76390-322 76390-150 76390-152 76390-154 76390-156 76390-158 76390-160 76390-162 76390-164 76390-166 76390-168 76390-178 76390-180 76390-182 76390-184 76390-194 76390-196 76390-198 76390-200 76390-202 76390-204 76390-206 76390-208 76390-210 76390-212 76390-222 76390-224 76390-226 76390-228 76390-230 76390-232 76390-234 76390-236 76390-246 76390-248 76390-250 76390-256 76390-258 76390-260 76390-262 76390-264 76390-274 76390-276 76390-278 76390-328 76390-330 76390-332 76390-334 76390-336 76390-346 76390-348 76390-350 76390-356 76390-358
Avantor® ACE® C18-300, HPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® C18-300 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
  • Designed for peptide and protein separations
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3, 5 and 10 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase - available on request
These columns provide excellent peak shape even at extremely low levels of additives such as TFA, making them ideal for LC/MS applications.