Avantor® ACE® Excel® NH₂, Analytical, HPLC/UHPLC Columns, 3 µm
Supplier: VWR
Avantor® ACE® Avantor® ACE® Excel®
EXL-1114-2502U EXL-1114-1203U EXL-1114-1546U EXL-1114-0546U EXL-1114-0246U EXL-1114-0303U EXL-1114-3546U EXL-1114-7503U EXL-1114-1246U EXL-1114-1502U EXL-1114-0203U EXL-1114-0302U EXL-1114-1046U ACE11140103GD EXL-1114-7546U EXL-1114-3502U EXL-1114-1202U EXL-1114-7502U EXL-1114-1002U EXL-1114-3503U EXL-1114-0503U EXL-1114-2503U EXL-1114-1503U EXL-1114-0502U EXL-1114-1003U EXL-1114-2046U EXL-1114-0346U ACE11140102GD EXL-1114-0202U EXL-1114-2546U
76393-326EA 2348.58 CAD
76393-326 76393-348 76393-328 76393-306 76393-308 76393-340 76393-320 76393-342 76393-344 76393-322 76393-324 76393-346 76393-360 76393-338 76393-316 76393-318 76384-498 76384-496 76393-330 76393-352 76393-332 76393-310 76393-354 76393-334 76393-312 76393-356 76393-314 76393-358 76393-336 76393-350
Avantor® ACE® Excel® NH₂, Analytical, HPLC/UHPLC Columns, 3 µm
Chromatography Columns
These stainless steel columns with proprietary bonding technology provide high robustness with excellent column lifetimes.
  • Usable in reversed phase, normal phase, weak anion-exchange and HILIC modes
  • Ideal for the analysis of sugars
  • Ultra-inert, high efficiency 1,7, 3 and 5 µm particles for optimum peak shape
  • Method validation kits contain three different batches of the same phase - available on request