Avantor® ACE® Advanced, Method Development Kits, 5 µm, HPLC/UHPLC Columns
Supplier: Avantor
MDKA-5-0502U MDKA-5-0503U MDKA-5-0546U MDKA-5-1002U MDKA-5-1003U MDKA-5-1046U MDKA-5-1502U MDKA-5-1503U MDKA-5-1546U MDKA-5-2502U MDKA-5-2503U MDKA-5-2546U
76396-394EA 1992.4 CAD
76396-394 76396-396 76396-398 76396-408 76396-410 76396-412 76396-436 76396-438 76396-440 76396-450 76396-452 76396-454
Avantor® ACE® Advanced, Method Development Kits, 5 µm, HPLC/UHPLC Columns
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1.7 and 5 µm ) and complementary phases
  • Analytical IDs in a
More Product Information