Avantor® ACE® CN, HPLC Columns, 10 µm
Supplier: HICHROM LIMITED
ACE-134-0110GD ACE-134-1246 ACE-134-1520 ACE-134-1539 ACE-134-2504 ACE-134-2546 ACE-134-3004
76388-924PK 414.61 CAD
76388-924 76388-926 76388-928 76388-930 76388-932 76388-934 76388-936 76388-938 76388-940 76388-942 76388-944 76388-946 76388-948 76388-950 76388-952 76388-958 76388-960 76388-962 76388-964 76388-966 76388-968 76388-970 76388-972 76388-974 76388-976 76388-978 76388-980 76388-982 76388-984 76388-986 76388-988 76388-990 76388-992 76388-994 76388-996 76388-998 76389-014 76389-016 76389-018
Avantor® ACE® CN, HPLC Columns, 10 µm
Chromatography Columns
These Avantor® ACE® CN columns provide great reproducibility and column lifetime.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Can be used under both reversed phase and normal phase conditions
  • Capillary to preparative dimensions
  • Wide range of particle sizes (2 to 10 µm) and complementary phases
  • Useful for the separation of polar analytes and compounds with double and triple bonds
  • Method validation kits contain three different batches of the same phase - available on request
These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative. These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.