Avantor® ACE® C18-Amide, HPLC/UHPLC Columns, 10 µm
Supplier: Avantor
ACE-1312-2510 ACE-1312-2520
76388-550PK 2327.83 CAD
76388-550 76388-554 76388-556 76388-558 76388-560 76388-562 76388-564 76388-566 76388-568 76388-570 76388-572 76388-578 76388-580 76388-582 76388-584 76388-586 76388-588 76388-590 76388-592 76388-594 76388-596 76388-598 76388-600 76388-602 76388-604 76388-626 76388-628 76388-630
Avantor® ACE® C18-Amide, HPLC/UHPLC Columns, 10 µm
Chromatography Columns
Avantor® ACE® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2.0 to 10 µm) and complementary phases
  • Low bleed for UV and LC/MS
More Product Information