Avantor® ACE® Bioanalytical, HPLC Columns, 300 Å Method Development Kits, 3 µm
Supplier: Avantor
MDKB-3-0502 MDKB-3-0503 MDKB-3-0546 MDKB-3-1002 MDKB-3-1003 MDKB-3-1046 MDKB-3-1502 MDKB-3-1503 MDKB-3-1546
76396-504EA 3596.05 CAD
76396-504 76396-506 76396-508 76396-518 76396-520 76396-522 76396-546 76396-548 76396-550
Avantor® ACE® Bioanalytical, HPLC Columns, 300 Å Method Development Kits, 3 µm
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (3 and 5 µm ) and complementary phases
  • Analytical IDs in a wide range of column lengths
  • Ideal for method development
  • Compatible with all manufacturers' LC systems
A complete range of kits for all requirements is available, including porous, solid core, HILIC and bioanalytical 300 Å column kits. These kits group together the essential column chemistries for method development and are available in a wide range of column formats.
Delivery information: Avantor® ACE® Bioanalytical Method Development Kits contain Avantor® ACE® C18-300, Avantor® ACE® C4-300 and Avantor® ACE® Phenyl-300 stainless steel columns.