Avantor® ACE® Generix® C8 & C8(2), Analytical HPLC Columns
Supplier: Avantor
GEN-3C8-0502 GEN-3C8-1046 GEN-3C8-1546 GEN-3C82-0502 GEN3C820502/3 GEN-3C82-0503 GEN-3C82-0504 GEN-3C82-0546 GEN-3C82-1002 GEN-3C82-1003 GEN-3C82-1004 GEN-3C82-1046 GEN3C821046/3 GEN-3C82-1202 GEN-3C82-1203 GEN-3C82-1204 GEN-3C82-1246 GEN-3C82-1502 GEN-3C82-1503 GEN-3C82-1504 GEN-3C82-1546 GEN3C821546/3 GEN-3C82-7502 GEN-3C82-7503 GEN-3C82-7504 GEN-3C82-7546 GEN-3C8-GD GEN-3C8-GD2 GEN-5C8-1046 GEN-5C8-1546 GEN-5C82-0502 GEN5C820502/3 GEN-5C82-0503 GEN-5C82-0504 GEN-5C82-0546 GEN-5C82-1002 GEN-5C82-1003 GEN-5C82-1004 GEN-5C82-1046 GEN-5C82-1202 GEN-5C82-1203 GEN-5C82-1204 GEN-5C82-1246 GEN-5C82-1502 GEN-5C82-1503 GEN-5C82-1504 GEN-5C82-1546 GEN5C821546/3 GEN-5C82-2502 GEN-5C82-2503 GEN-5C82-2504 GEN-5C82-2546 GEN5C822546/3 GEN-5C8-2546 GEN-5C82-7502 GEN-5C82-7503 GEN-5C82-7504 GEN-5C82-7546 GEN-5C8-GD GEN-C82-GD GEN-C82-GD2
76394-796EA 953.26 CAD
76394-796 76394-798 76394-800 76394-802 76394-804 76394-806 76394-808 76394-810 76394-812 76394-814 76394-816 76394-818 76394-820 76394-822 76394-824 76394-826 76394-828 76394-830 76394-832 76394-834 76394-836 76394-838 76394-840 76394-842 76394-844 76394-846 76394-848 76394-850 76395-010 76395-012 76395-014 76395-016 76395-018 76395-020 76395-022 76395-024 76395-026 76395-028 76395-030 76395-032 76395-034 76395-036 76395-038 76395-040 76395-042 76395-044 76395-046 76395-048 76395-050 76395-052 76395-054 76395-056 76395-058 76395-064 76395-066 76395-068 76395-070 76395-072 76395-074 76395-198 76395-200
Avantor® ACE® Generix® C8 & C8(2), Analytical HPLC Columns
Chromatography Columns
These high performance HPLC columns contain high purity spherical silica.
  • Ultra-narrow particle size distribution for excellent column efficiency
  • Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
  • Excellent column to column and batch to batch reproducibility
  • These columns are made of stainless steel
Each column is individually tested and supplied with a test chromatogram.
Two generations of Avantor® ACE® Generix® C8 exist: C8 and C8(2) they differ in pore size, carbon load and surface area.