Avantor® ACE® Bioanalytical, HPLC Columns, 300 Å Method Development Kits, 5 µm
Supplier: Avantor
MDKB-5-0502 MDKB-5-0503 MDKB-5-0546 MDKB-5-1002 MDKB-5-1003 MDKB-5-1046 MDKB-5-1502 MDKB-5-1503 MDKB-5-1546 MDKB-5-2502 MDKB-5-2503 MDKB-5-2546
76396-600EA 2163.59 CAD
76396-600 76396-602 76396-604 76396-614 76396-616 76396-618 76396-642 76396-644 76396-646 76396-656 76396-658 76396-660
Avantor® ACE® Bioanalytical, HPLC Columns, 300 Å Method Development Kits, 5 µm
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (3 and 5 µm) and complementary phases
  • Analytical IDs in a wide
More Product Information