Avantor® ACE® C18-AR, HPLC Columns, 10 µm
Supplier: Avantor
ACE-139-1546 ACE-139-2510 ACE-139-2546
76389-306PK 429.73 CAD
76389-306 76389-308 76389-310 76389-312 76389-314 76389-316 76389-318 76389-320 76389-322 76389-324 76389-326 76389-328 76389-330 76389-332 76389-334 76389-340 76389-342 76389-344 76389-346 76389-348 76389-350 76389-352 76389-354 76389-356 76389-358 76389-360 76389-362 76389-364 76389-366 76389-368 76389-370 76389-372 76389-374 76389-376 76389-378 76389-380 76389-396 76389-398 76389-400
Avantor® ACE® C18-AR, HPLC Columns, 10 µm
Chromatography Columns
The Avantor® ACE® C18-AR phase utilises a specially developed ligand combining a C18 chain with integral phenyl functionality, thus combining the benefits of both C18 and phenyl characteristics into a single phase. This can be used for ‘standard’ C18 column separations, but is additionally recommended for separations that involve compounds containing aromatic functionality.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2 to 10 µm) and
More Product Information