Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 2 µm
Supplier: Avantor
MDKE-2-0502U MDKE-2-0503U MDKE-2-0546U MDKE-2-1002U MDKE-2-1003U MDKE-2-1046U MDKE-2-1502U MDKE-2-1503U MDKE-2-1546U
76396-848EA 3017.69 CAD
76396-848 76396-850 76396-852 76396-862 76396-864 76396-866 76396-888 76396-890 76396-892
Avantor® ACE®, HPLC/UHPLC Columns, Extended Method Development Kits, 2 µm
Chromatography Columns
Avantor® ACE® Method Development Kits (MDK) are designed to maximise selectivity, offering a powerful and reliable approach to UHPLC/HPLC method development. Based upon an ultra-inert, high efficiency silica, Avantor® ACE® phases incorporate the latest developments in LC stationary phase design, providing chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
  • Porous, solid core, HILIC and biomolecule kit options
  • Wide range of particle sizes (1.7 and 5 µm ) and complementary phases
  • Analytical IDs in a
More Product Information