Avantor® ACE® C18-HL, HPLC Columns, 10 µm
Supplier: Avantor
ACE-331-0103GD ACE-331-0110GD ACE-331-1010 ACE-331-2510 ACE-331-2546
76391-880PK 1978.22 CAD
76391-880 76391-882 76391-884 76391-886 76391-888 76391-890 76391-892 76391-894 76391-896 76391-898 76391-900 76391-902 76391-904 76391-906 76391-908 76391-914 76391-916 76391-918 76391-920 76391-922 76391-924 76391-926 76391-928 76391-930 76391-932 76391-934 76391-936 76391-938 76391-940 76391-942 76391-944 76391-946 76391-948 76391-950 76391-966 76391-968 76391-970
Avantor® ACE® C18-HL, HPLC Columns, 10 µm
Chromatography Columns
Avantor® ACE® C18-HL columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (3 to 15 µm) and complementary phases
  • Especially suited for preparative work due to higher carbon load
  • Method validation kits contain three different batches of the same phase - available on
More Product Information