Avantor® ACE® Generix® C18 & C18(2), Analytical HPLC Columns
Supplier: VWR
Avantor® ACE® Generix®
GEN-5C182-2546 GEN-5C182-1203 GEN-5C18-1046 GEN-5C182-0503 GEN-3C182-7503 GEN-3C18-1002 GEN3C1821046/3 GEN-3C182-1202 GEN-5C182-1002 GEN3C1820502/3 GEN-5C182-1502 GEN-3C182-1002 GEN-5C182-2503 GEN-3C182-0504 GEN-5C182-1204 GEN5C1821546/3 GEN-3C182-1203 GEN-5C182-7503 GEN-3C182-7502 GEN-5C18-2546 GEN-5C182-1504 GEN5C1820502/3 GEN-3C182-1004 GEN-3C182-0503 GEN3C1821546/3 GEN-5C182-0502 GEN-3C18-1046 GEN-3C182-0502 GEN-3C182-1204 GEN-3C18-0502 GEN-5C182-2502 GEN5C1822546/3 GEN-5C182-0546 GEN-3C182-0546 GEN-5C182-7546 GEN-5C182-1046 GEN-5C182-2504 GEN-5C182-0504 GEN-3C182-1502 GEN-3C18-1502 GEN-3C182-1003 GEN-5C182-1546 GEN-3C182-7504 GEN-C182-GD GEN-5C18-1546 GEN-3C182-1546 GEN-3C182-7546 GEN-5C182-1004 GEN-3C18-GD GEN-5C182-3046 GEN-5C182-1003 GEN-5C182-1202 GEN-3C182-1046 GEN-C182-GD2 GEN-3C182-1504 GEN-3C182-1246 GEN-5C182-7502 GEN-5C182-1503 GEN-3C18-1546 GEN-5C182-3004 GEN-3C182-1503 GEN-5C182-1246 GEN-5C18-GD GEN-5C182-7504
76394-774EA 761.04 CAD
76394-774 76394-972 76394-970 76394-772 76394-770 76394-738 76394-978 76394-976 76394-778 76394-974 76394-776 76394-786 76394-742 76394-984 76394-740 76394-982 76394-784 76394-980 76394-782 76394-780 76394-948 76394-748 76394-946 76394-944 76394-988 76394-746 76394-788 76394-986 76394-744 76394-950 76394-752 76394-994 76394-750 76394-992 76394-794 76394-990 76394-792 76394-790 76395-192 76394-958 76395-194 76394-758 76394-956 76394-998 76394-756 76394-954 76394-952 76394-754 76394-996 76395-002 76395-004 76395-000 76395-006 76395-008 76394-962 76394-764 76394-762 76394-960 76394-760 76394-968 76394-768 76394-966 76394-766 76394-964
Avantor® ACE® Generix® C18 & C18(2), Analytical HPLC Columns
Chromatography Columns
These high performance HPLC columns contain high purity spherical silica.
  • Ultra-narrow particle size distribution for excellent column efficiency
  • Sharp, highly symmetrical peaks for acidic, basic and neutral analytes
  • Excellent column to column and batch to batch reproducibility
Each column is individually tested and supplied with a test chromatogram.
Two generations of Avantor® ACE® Generix® C18 exist: C18 and C18(2) they differ in pore size, carbon load and surface area.