Writable Cryogenic Multi-Color Labels for Microtubes Tops
Supplier: GA INTERNATIONAL INC
807656068077 807656068053 807656067940 807656068084 807656068008 807656067971 807656068015 807656067957 807656068022 807656067964 807656068046 807656068060
76275-686PK 32.56 CAD
76275-686 76275-688 76275-690 76275-692 76275-694 76275-696 76275-698 76275-704 76275-706 76275-708 76275-710 76275-712
Writable Cryogenic Multi-Color Labels for Microtubes Tops
Labels
Writable cryo color dots and rectangles for identifying the sides and tops of cryo vials and microtubes stored in lab freezers and liquid nitrogen dewars. Easy to write on with permanent cryo markers. JTA-class labels are available in a variety of colors.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Available in a range of colors for color-coding and easy identification
  • Writable rectangular labels for deep-freeze storage
  • Strong adhesive ensures labels won’t wrinkle or peel during long-term storage in freezers and on dry ice
Ideal for identification of vials, plastic bags, and other containers for long-term storage and color-coding in deep-freezing and harsh laboratory environments.