Avantor® ACE® C18-Amide, HPLC/UHPLC Columns, 5 µm
Supplier: Avantor
ACE-1212-1520 ACE-1212-2504 ACE-1212-2510 ACE-1212-2520 ACE-12120103GD ACE-12120110GD ACE12120120GD ACE-1212-0520 ACE-1212-1010 ACE-1212-1020 ACE-1212-1030
76386-246EA 6058.76 CAD
76386-246 76386-248 76386-250 76386-252 76386-254 76386-256 76386-258 76386-260 76386-262 76386-264 76386-266 76386-268 76386-270 76386-272 76386-274 76386-276 76386-278 76386-280 76386-282 76386-332 76386-334 76386-182 76386-184 76386-186 76386-188 76386-190 76386-192 76386-194 76386-196 76386-200 76386-202 76386-204 76386-206 76386-208 76386-210 76386-212 76386-214 76386-216 76386-218 76386-220 76386-222 76386-224 76386-226 76386-228 76386-230 76386-232 76386-234 76386-236 76386-238 76386-240 76386-284 76386-320 76386-322 76386-324 76386-326
Avantor® ACE® C18-Amide, HPLC/UHPLC Columns, 5 µm
Chromatography Columns
Avantor® ACE® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (2.0 to 10 µm) and complementary phases
  • Low bleed for UV and LC/MS
More Product Information