Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
EXL-1211-0202U EXL-1211-0302U EXL-1211-0346U EXL-1211-0502U EXL-1211-0546U EXL-1211-1002U EXL-1211-1046U EXL-1211-1246U EXL-1211-1502U EXL-1211-1503U EXL-1211-1546U EXL-1211-2046U EXL-1211-2503U EXL-1211-2546U EXL-1211-3546U EXL-1211-7503U EXL-1211-7546U
76393-816EA 2029.51 CAD
76393-816 76393-818 76393-820 76393-822 76393-824 76393-826 76393-830 76393-832 76393-834 76393-836 76393-838 76393-840 76393-842 76393-844 76393-846 76393-848 76393-850 76393-852 76393-858 76393-862 76393-864 76393-866 76393-868 76393-870 76393-872 76393-880 76393-882 76393-884
Avantor® ACE® Excel® SuperC18, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
Avantor® ACE® Excel® SuperC18™ columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
  • Ultra-inert for maximum performance and reproducibility
  • Ultra low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1.7 to 10 µm) and complementary phases
These columns are made of stainless steel.