Autoclave-Resistant Labels for Inkjet Printers
Supplier: GA INTERNATIONAL INC
807656009643 807656009650 807656009667
76272-012PK 170.67 CAD
76272-012 76272-022 76272-024
Autoclave-Resistant Labels for Inkjet Printers
Labels
Autoclave-resistant labels for desktop inkjet printers. Can be printed with eye-catching, full-color logos and graphics. These permanent inkjet labels will adhere to different surfaces for identifying various sterilization containers and materials.
  • Temperature range: −40 to +150 °C (−40 to +302 °F)
  • Autoclavable labels withstand ten cycles in steam autoclaves and dry heat ovens
  • Strong permanent adhesive adheres to glassware, metal, and plastic surfaces
  • Compatible with all major brands of desktop inkjet printers
Permanent inkjet printable labels areMore Product Information