Clear Thermal-Transfer Labels for Delicate Surfaces
Supplier: GA INTERNATIONAL INC
807656033082 807656033099 807656033068 807656033075 807656033044 807656033051
76210-134RL 96.47 CAD
76210-134 76210-180 76210-334 76210-336 76210-338 76210-340
Clear Thermal-Transfer Labels for Delicate Surfaces
Labels
These transparent labels will adhere to a variety of smooth, untextured, and delicate surfaces. They can easily be re-positioned with virtually no residue left behind upon removal. Printable with thermal transfer printers for a 'printed-on look'.
  • Temperature range: −45 to +82 °C (−49 to +179.6 °F)
  • Labels adhere using static cling-like properties, ensuring virtually no adhesive residue is left behind upon removal
  • Transparent design offers a clean, modern, and 'printed-on' look
  • Printable with all major thermal transfer printer brands
Transparent labels affix to a variety of smooth, untextured surfaces. They can easily be re-positioned on flat and gently curved surfaces, making them perfect for temporarily labeling delicate surfaces. Ideal for identifying most non-frosted glass surfaces such as labware, pyrex bottles, windows, aquariums, terrariums, and eyeglass lenses.
These labels are also suitable for identifying smooth surfaces, including bags, coated paper and carton, galvanized, epoxy coated, painted, and stainless steel, laminated wood products, painted drywall, polished, ceramic, porcelain surfaces and tiles, chrome, synthetic, natural, and composite countertops, melamine, silicone, teflon, epoxy resin, polished carbon fiber, and other similar non-porous glossy surfaces. Printable with thermal transfer printers, they are also writable with permanent markers and ballpoint pens.