Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
ACE-112-0102GD ACE-112-0103GD EXL-112-0502U EXL-112-0503U EXL-112-0546U EXL-112-1002U EXL-112-1003U EXL-112-1046U EXL-112-1203U EXL-112-1246U EXL-112-1502U EXL-112-1503U EXL-112-1546U EXL-112-2546U EXL-112-7503U EXL-112-7546U
76384-572PK 490.18 CAD
76384-572 76384-574 76393-368 76393-370 76393-372 76393-374 76393-376 76393-378 76393-380 76393-382 76393-384 76393-386 76393-388 76393-390 76393-392 76393-394 76393-396 76393-398 76393-400 76393-402 76393-404 76393-406 76393-408 76393-410 76393-412 76393-414 76393-416 76393-418 76393-420
Avantor® ACE® Excel® C8, HPLC/UHPLC Columns, Analytical, 3 µm
Chromatography Columns
These ultra-inert Avantor® ACE® Excel® C8 columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Can be used as general purpose starting point column
  • Wide range of particle sizes (2, 3 and 5 µm) and complementary phases
These columns are designed for a wide range of chromatographic applications, to provide great performance with acidic, basic and neutral molecules.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.