Cryogenic Tapes for Laboratory Use
Supplier: GA INTERNATIONAL INC
807656042329 807656042336 807656042343 807656042350 807656042367 807656042374 807656079745 807656079738 807656079752 807656079707 807656079721 807656079714 807656079806 807656079790 807656079813 807656079769 807656079783 807656079776 807656079868 807656079851 807656079875 807656079820 807656079844 807656079837
76275-356RL 57.01 CAD
76275-356 76275-854 76275-856 76275-858 76275-860 76275-862 76275-864 76275-866 76275-868 76275-870 76275-872 76275-874 76275-876 76275-878 76275-880 76275-882 76275-884 76275-886 76275-888 76275-890 76275-892 76275-894 76275-896 76275-898
Cryogenic Tapes for Laboratory Use
Tapes
This water-proof cryogenic tape is designed for identifying cryogenic containers stored long-term under cryo conditions, including in liquid nitrogen tanks.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • For use in harsh cryogenic conditions
  • Cryogenic permanent adhesive ensures tape won’t peel or crack when exposed to liquid nitrogen
  • Continuous tape format allows use of as much or little as necessary
Continuous writable tape for cryogenic use. Water-proof, resistant to liquid nitrogen, dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments. Featuring an extra strong adhesive, this tape is available in a variety of colors for freezer and liquid nitrogen use (as low as −196 °C) on plastic and glass containers.