Cryogenic Color Dots Writable Labels
Supplier: GA INTERNATIONAL INC
807656024905 807656024912 807656024929 807656024936 807656024943 807656000060 807656024950 807656024967 807656024974 807656024981 807656024998 807656025001 807656024899 807656025018 807656025025 807656025032 807656003498 807656025049 807656025056 807656025063 807656025070 807656025087 807656025094 807656025100 807656025117 807656025124 807656025131 807656025148 807656025155 807656025162 807656025179 807656025186 807656025193 807656025209 807656000336 807656000473 807656000343 807656000350 807656000367 807656000374 807656000381 807656000398 807656000404 807656000411 807656000428 807656000435 807656000442 807656000459 807656000466 807656000480 807656025230 807656025247 807656025254 807656025261 807656025278 807656025285
76275-326PK 26.26 CAD
76275-326 76275-328 76275-330 76275-332 76275-334 76275-336 76275-338 76275-340 76275-446 76275-448 76275-450 76275-452 76275-596 76275-598 76275-600 76275-602 76275-604 76275-606 76275-608 76275-610 76275-612 76275-614 76275-616 76275-618 76275-620 76275-622 76275-624 76275-626 76275-628 76275-630 76275-632 76275-634 76275-636 76275-638 76275-640 76275-642 76275-644 76275-646 76275-648 76275-650 76275-652 76275-654 76275-656 76275-658 76275-660 76275-662 76275-664 76275-666 76275-668 76275-670 76275-674 76275-676 76275-678 76275-680 76275-682 76275-684
Cryogenic Color Dots Writable Labels
Labels
These small diameter, round cryo labels for microtube tops are made with a strong adhesive. Ideal for −80 °C freezers, dry ice, and liquid nitrogen storage.
  • Temperature range: −196 to +110 °C (−321 to +230 °F)
  • Available in a wide range of colors, for color-coding and easy identification
  • Easy to write on with permanent markers
  • Small round labels are ideal for 0.5 to 1.5 ml Eppendorf tube caps and vials
  • Strong adhesive ensures labels won't wrinkle or peel during long-term storage
  • Resistant to gamma irradiation (tested to up to 50 kGy)
Specially designedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable