Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 3 µm
Supplier: Avantor
EXL-1112-0246U EXL-1112-0302U EXL-1112-0503U EXL-1112-1002U EXL-1112-1003U EXL-1112-1046U EXL-1112-1202U EXL-1112-1502U EXL-1112-1503U EXL-1112-1546U EXL-1112-2546U EXL-1112-3546U EXL-1112-7502U EXL-1112-7503U EXL-1112-7546U ACE-11120102GD ACE-11120103GD
76393-182EA 1276.72 CAD
76393-182 76393-184 76393-186 76393-188 76393-190 76393-192 76393-194 76393-196 76393-198 76393-200 76393-202 76393-204 76393-206 76393-208 76393-210 76393-212 76393-214 76393-216 76393-218 76393-220 76393-222 76393-224 76393-226 76393-228 76393-230 76393-238 76393-240 76393-242 76384-358 76384-360
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 3 µm
Chromatography Columns
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS compatibility
  • C18 hydrophobic plus Amide interactions, compatible with 100% aqueous condition
The special design of the Avantor® ACE® C18-Amide ligand incorporates an extended alkyl chain spacer between the silica surface and amide polar group, which gives extra phase stability.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.