Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
ACE-12130103GD EXL-1213-0502U EXL-1213-1002U EXL-1213-1046U EXL-1213-1546U EXL-1213-2546U
76386-352PK 414.78 CAD
76386-352 76386-354 76393-948 76393-950 76393-952 76393-954 76393-956 76393-958 76393-960 76393-962 76393-964 76393-966 76393-968 76393-970 76393-972 76393-974 76393-976 76393-978 76393-980 76393-982 76393-984 76393-986 76393-988 76393-990 76393-992 76393-994 76393-996 76394-000 76394-002 76394-004
Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
Avantor® ACE® CN-ES combines CN polar selectivity with enhanced hydrophobicity, enabling the benefits of both interactions to be fully exploited. Extra phase stability and increased column lifetime is provided by an extended alkyl chain spacer between the silica surface and cyano group. Avantor® ACE® CN-ES is suitable for both reversed phase and normal phase separations.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • With hydrophobic plus CN polar selectivity, especially useful for polar molecules
  • Wide range of particle sizes (1
More Product Information