Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: HICHROM LIMITED
EXL-1212-0502U EXL-1212-0546U EXL-1212-1002U EXL-1212-1046U EXL-1212-1502U EXL-1212-1503U EXL-1212-1546U EXL-1212-2502U EXL-1212-2503U EXL-1212-2546U
76393-886EA 1206.24 CAD
76393-886 76393-888 76393-890 76393-892 76393-894 76393-896 76393-898 76393-900 76393-902 76393-904 76393-906 76393-908 76393-910 76393-912 76393-914 76393-916 76393-918 76393-920 76393-922 76393-924 76393-926 76393-928 76393-930 76393-932 76393-934 76393-942 76393-944 76393-946
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS compatibility
  • C18 hydrophobic plus Amide interactions, compatible with 100% aqueous condition
The special design of the Avantor® ACE® C18-Amide ligand incorporates an extended alkyl chain spacer between the silica surface and amide polar group, which gives extra phase stability.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.