Avantor® ACE® Excel® C18-Amide, UHPLC Columns, 1.7 µm
Supplier: Avantor
EXL-1712-0202U EXL-1712-0302U EXL-1712-0502U EXL-1712-0503U EXL-1712-1002U EXL-1712-1003U EXL-1712-7502U
76394-464EA 2656.52 CAD
76394-464 76394-466 76394-468 76394-470 76394-472 76394-474 76394-476 76394-478 76394-480 76394-482 76394-484 76394-486
Avantor® ACE® Excel® C18-Amide, UHPLC Columns, 1.7 µm
Chromatography Columns
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS compatibility
  • C18 hydrophobic plus Amide interactions, compatible with 100% aqueous condition
The special design of the Avantor® ACE® C18-Amide ligand incorporates an extended alkyl chain spacer between the silica surface and amide polar group, which gives extra phase stability.
Ordering information: Method validation kits contain three different batches of the same phase - available on request.