Avantor® ACE® SuperC18, HPLC Columns, 3 µm

Supplier: Avantor
ACE-1111-75005 ACE-1111-10005
76384-256EA 1722.04 CAD
76384-256 76384-258 76384-260 76384-262 76384-268 76384-270 76384-272 76384-274 76384-346 76384-348 76384-350 76384-352 76384-354 76384-276 76384-278 76384-280 76384-282 76384-284 76384-286 76384-288 76384-290 76384-294 76384-296 76384-298 76384-300
Avantor® ACE® SuperC18, HPLC Columns, 3 µm
Chromatography Columns
Avantor® ACE® SuperC18™ columns provide high stability across an extended pH range. These stainless-steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.

  • Ultra-inert for maximum performance and reproducibility
  • Ultra-low bleed for improved LC/MS compatibility
  • Wide range of particle sizes (1.7 to 10 µm) and complementary phases

Maximum recommended pH: 1.5 to 11.5

Max. pressure: 275 bar (3989 psi).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR