Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, 2 µm
Supplier: Avantor
EXL-1012-0502U EXL-1012-0503U EXL-1012-0546U EXL-1012-1002U EXL-1012-1003U EXL-1012-1046U EXL-1012-1202U EXL-1012-1203U EXL-1012-1502U EXL-1012-1503U EXL-1012-1546U EXL-1012-7502U EXL-1012-7503U EXL-1012-7546U ACE-1P12-GD2U ACE-1P12-GD4U
76392-600EA 1361.74 CAD
76392-600 76392-602 76392-604 76392-606 76392-608 76392-610 76392-612 76392-614 76392-616 76392-618 76392-620 76392-622 76392-624 76392-626 76392-628 76392-630 76392-632 76392-634 76392-636 76392-638 76392-640 76392-642 76392-644 76392-646 76408-406 76408-408
Avantor® ACE® Excel® C18-Amide, HPLC/UHPLC Columns, 2 µm
Chromatography Columns
Avantor® ACE® Excel® C18-Amide is a uniquely designed stationary phase offering improved retention of polar compounds, enhanced phenolic resolution and alternative selectivity to C18 bonded columns. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra-high purity, base deactivated silica with exceptional reproducibility
  • Ideal for a wide range of analytes differing in hydrophobicity
  • Wide range of particle sizes (1.7 to 5 µm) and complementary phases
  • Low bleed for UV and LC/MS
More Product Information