Avantor® ACE® Excel® Phenyl, HPLC/UHPLC Columns, Analytical, 5 µm
Supplier: Avantor
EXL-125-0302U EXL-125-0502U EXL-125-1002U EXL-125-1003U EXL-125-1046U EXL-125-1502U EXL-125-1546U EXL-125-2546U
76394-210EA 1242.04 CAD
76394-210 76394-212 76394-214 76394-216 76394-218 76394-220 76394-222 76394-224 76394-226 76394-228 76394-230 76394-232 76394-234 76394-236 76394-238 76394-240 76394-242 76394-244 76394-246 76394-248 76394-250 76394-252 76394-254 76394-256
Avantor® ACE® Excel® Phenyl, HPLC/UHPLC Columns, Analytical, 5 µm
Chromatography Columns
These Avantor® ACE® Excel® Phenyl columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Offers strong π-π interactions
  • Ideal for analytes with π-bonding, electron withdrawing or delocalized groups
  • Wide range of particle sizes available (2, 3 and 5 µm) and complementary phases
  • Method validation kits contain three different batches of the same phase -
More Product Information