Avantor® ACE® Excel® AQ, Analytical HPLC/UHPLC Columns, 2 µm
Supplier: Avantor
EXL-106-0502U EXL-106-0503U EXL-106-0546U EXL-106-1002U EXL-106-1003U EXL-106-1046U EXL-106-1502U EXL-106-1503U EXL-106-7502U ACE-1P6-GD2U ACE-1P6-GD4U
76392-898EA 1687.6 CAD
76392-898 76392-900 76392-902 76392-904 76392-906 76392-908 76392-910 76392-912 76392-914 76392-916 76392-918 76392-920 76392-922 76392-924 76392-926 76392-928 76392-930 76392-932 76392-934 76392-936 76392-938 76392-940 76392-942 76392-944 76408-430 76408-432
Avantor® ACE® Excel® AQ, Analytical HPLC/UHPLC Columns, 2 µm
Chromatography Columns
Avantor® ACE® Excel® AQ columns provide great reproducibility and column lifetime. These stainless steel columns are available in a wide range of particle sizes and dimensions from capillary to preparative.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • Retains polar compounds in 100% aqueous mobile phase
  • Unique C18 bonded phase with integral polar functionality
  • Ideal for use with fast gradients due to its rapid re-equilibration properties and resistance to retention loss
  • Wide range of particle sizes (2 to 5 µm) and
More Product Information