Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 1.7 µm
Supplier: Avantor
EXL-1713-0502U EXL-1713-0503U EXL-1713-1002U EXL-1713-1003U EXL-1713-7502U EXL-1713-7503U
76394-488EA 1918.82 CAD
76394-488 76394-490 76394-492 76394-494 76394-496 76394-498 76394-500 76394-502 76394-504 76394-506 76394-512 76394-514
Avantor® ACE® Excel® CN-ES, HPLC/UHPLC Columns, Analytical, 1.7 µm
Chromatography Columns
Avantor® ACE® CN-ES combines CN polar selectivity with enhanced hydrophobicity, enabling the benefits of both interactions to be fully exploited. Extra phase stability and increased column lifetime is provided by an extended alkyl chain spacer between the silica surface and cyano group. Avantor® ACE® CN-ES is suitable for both reversed phase and normal phase separations.
  • Ultra high purity, base deactivated silica with exceptional reproducibility
  • With hydrophobic plus CN polar selectivity, especially useful for polar molecules
  • Wide range of particle sizes (1
More Product Information